Skip to Content
Merck
All Photos(4)

Key Documents

922005

Sigma-Aldrich

Droplet generator chip - Four elements per chip design

Fluidic 537, COC

Synonym(s):

Microfluidic chip, Microparticle synthesis, Nanoparticle

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$170.00

$170.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$170.00

About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

$170.00


Check Cart for Availability

Request a Bulk Order

description

Microfludic chip x1

Looking for similar products? Visit Product Comparison Guide

Application

Microfluidic generation of droplets can produce highly monodispersed droplets with high frequency (up to hundreds of kHz). Interest in droplet-based microfluidic systems has grown substantially, because microfluidics offers the ability to handle very small volumes (μl to fl) of fluids, provides better mixing, encapsulation, sorting, and sensing. Microfluidics can be used for high throughput experimentation. Microfluidic-based droplets have many diverse and varied applications such as particle synthesis and chemical analysis. Highly controlled droplet production also makes single cell analysis, or drug testing possible.

Droplet generator chip - Four elements per chip design, Fluidic 537, COC is made of COC (Cyclic olefin copolymer ). The droplet generator with design number 537 possesses a classic flow focusing geometry, suited to generate simple droplets. With four identical droplet generator units on one microfluidic chip, this device is ideal to compare multiple experimental setups in parallel, without the need of ordering a new chip for every experiment. Please be aware that this chip is one of the few droplet generators devices with Luer interfaces and appropriate Luer compatible accessories are required.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances of controlled drug delivery using microfluidic platforms
Li X, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2018)
Microfluidic-assisted fabrication of carriers for controlled drug delivery.
Santos H A, et al.
Lab on a chip, 17, 1856-1883 (2017)
Recent advances of controlled drug delivery usingmicrofluidic platforms.
Li X, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2018)

Articles

Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service