Skip to Content
Merck
All Photos(1)

Key Documents

920908

Sigma-Aldrich

(S,R,S)-VL285 Phenol

Synonym(s):

(2S,4R)-4-Hydroxy-N-(2-hydroxy-4-(4-methylthiazol-5-yl)benzyl)-1-((S)-3-methyl-2-(1-oxoisoindolin-2-yl)butanoyl)pyrrolidine-2-carboxamide, VHL E3 ligase ligand for PROTAC® research

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$847.00

$847.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$847.00

About This Item

Empirical Formula (Hill Notation):
C29H32N4O5S
CAS Number:
Molecular Weight:
548.65
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$847.00


Check Cart for Availability

Request a Bulk Order

ligand

VL285 phenol

Quality Level

form

solid

reaction suitability

reagent type: ligand

functional group

amine

storage temp.

2-8°C

SMILES string

O=C([C@@H]1C[C@@H](O)CN1C([C@H](C(C)C)N2CC(C=CC=C3)=C3C2=O)=O)NCC4=CC=C(C5=C(C)N=CS5)C=C4O

InChI

1S/C29H32N4O5S/c1-16(2)25(33-13-20-6-4-5-7-22(20)28(33)37)29(38)32-14-21(34)11-23(32)27(36)30-12-19-9-8-18(10-24(19)35)26-17(3)31-15-39-26/h4-10,15-16,21,23,25,34-35H,11-14H2,1-3H3,(H,30,36)/t21-,23+,25+/m1/s1

InChI key

KNZQRBRUYSXXRG-VTZPFEBOSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saul Jaime-Figueroa et al.
Bioorganic & medicinal chemistry letters, 30(3), 126877-126877 (2019-12-28)
A new series of Proteolysis Targeting Chimeras (PROTACs) targeting Bruton's Tyrosine Kinase (BTK) was synthesized, with the goal of improving the pharmacokinetic properties of our previously reported PROTAC, MT802. We recently described the ability of MT802 to induce degradation of
Christian Steinebach et al.
Chemical science, 11(13), 3474-3486 (2020-11-03)
Cyclin-dependent kinase 6 (CDK6) is an important regulator of the cell cycle. Together with CDK4, it phosphorylates and inactivates retinoblastoma (Rb) protein. In tumour cells, CDK6 is frequently upregulated and CDK4/6 kinase inhibitors like palbociclib possess high activity in breast
Blake E Smith et al.
Nature communications, 10(1), 131-131 (2019-01-12)
PROteolysis-TArgeting Chimeras (PROTACs) are hetero-bifunctional molecules that recruit an E3 ubiquitin ligase to a given substrate protein resulting in its targeted degradation. Many potent PROTACs with specificity for dissimilar targets have been developed; however, the factors governing degradation selectivity within
Dennis L Buckley et al.
ACS chemical biology, 10(8), 1831-1837 (2015-06-13)
Small molecule-induced protein degradation is an attractive strategy for the development of chemical probes. One method for inducing targeted protein degradation involves the use of PROTACs, heterobifunctional molecules that can recruit specific E3 ligases to a desired protein of interest.

Articles

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service