Skip to Content
Merck
All Photos(1)

Key Documents

919616

Sigma-Aldrich

[Pd(IPr#)(cin)Cl]

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$264.00
500 MG
$802.00

$264.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$264.00
500 MG
$802.00

About This Item

Empirical Formula (Hill Notation):
C102H81ClN2Pd
Molecular Weight:
1476.62
UNSPSC Code:
12352103
NACRES:
NA.22

$264.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

[Pd(IPr#)(cin)Cl] is a Pd-catalyst that can be used in a broad range of reactions, including cross-coupling, C-H functionalization, electrophilic additions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service