Skip to Content
Merck
All Photos(3)

Key Documents

919365

Sigma-Aldrich

(MeBPI)Ni-OAc

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$507.00

$507.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$507.00

About This Item

Empirical Formula (Hill Notation):
C22H19N5NiO2
Molecular Weight:
444.11
UNSPSC Code:
12352200
NACRES:
NA.22

$507.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

(MeBPI)Ni-OAc is a Ni catalyst developed in the Sevov lab for electrocatalytic cross-electrophile coupling reactions. When combined with a redox-active shuttle (918105), (MeBPI)Ni-OAc enables the reductive coupling of a wide range of aryl, heteroaryl, or vinyl bromides with primary or secondary alkyl bromides.

Product can be used with our Synlectro electrochemistry platform (ESYNTH001)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blaise L Truesdell et al.
Journal of the American Chemical Society, 142(12), 5884-5893 (2020-03-03)
Cross-electrophile coupling (XEC) of alkyl and aryl halides promoted by electrochemistry represents an attractive alternative to conventional methods that require stoichiometric quantities of high-energy reductants. Most importantly, electroreduction can readily exceed the reducing potentials of chemical reductants to activate catalysts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service