Skip to Content
Merck
All Photos(1)

Key Documents

919241

Sigma-Aldrich

Aluminum cerium oxide

nanopowder, ≤120 nm particle size (BET), 99% trace metals basis

Synonym(s):

Cerium aluminate, Cerium aluminum oxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$215.00
50 G
$694.00

$215.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$215.00
50 G
$694.00

About This Item

Linear Formula:
AlCeO3
CAS Number:
Molecular Weight:
215.10
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.23

$215.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99% trace metals basis

form

nanopowder

particle size

≤120 nm (BET)

SMILES string

[Ce+3].[O-][Al]([O-])[O-]

InChI

1S/Al.Ce.3O/q;+3;3*-1

InChI key

GFIRVRHWPCRRBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum cerium oxide finds applications in in defense, automotive, aerospace and electrical industries.

Packaging

10, 50 g in glass bottle

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service