Skip to Content
Merck
All Photos(1)

Key Documents

917311

Sigma-Aldrich

Indium tin oxide coated PEN

with 0% transparent Pt, L × W × thickness 9.6 cm × 9.6 cm × 125 μm, sheet resistance 5 Ω/sq

Synonym(s):

ITO coated polyethylene naphthalate with sputtered Pt layer, ITO-PEN with sputtered Pt layer, PECF-CAT

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$409.00

$409.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$409.00

About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

$409.00


Check Cart for Availability

Request a Bulk Order

material

substrate (Polyethylene naphlalate (PEN) film (one side coated))

Quality Level

description

with 0% transparent Pt
(Catalyst material: Pt/Ti alloy)

sheet resistance

5 Ω/sq

L × W × thickness

9.6 cm × 9.6 cm × 125 μm

transmittance

0% (non-transparent)

Application

This product is an one-side coated, flexible, ITO coated PEN polymer film. It is a ready-to-use flexible substrate with low sheet resistance. Pt catalyste was sputtered onto the coated side of the film with 0% transmittance.
This product can be used for rapid fabrication of flexible Perovskite solar cells and flexible dye-sensitized solar cells.[1][2][3]

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photovoltaic Performance of Plastic Dye-Sensitized Electrodes Prepared by Low-Temperature Binder-Free Coating of Mesoscopic Titania
Miyasaka T, et al.
Journal of the Electrochemical Society, 154 (2007)
Jennifer M Pringle et al.
Chemical communications (Cambridge, England), 46(29), 5367-5369 (2010-06-23)
Electrodeposition of poly(3,4-ethylenedioxythiophene) onto conducting plastic allows the cheap and facile synthesis of plastic cathodes for dye-sensitised solar cells, having excellent solar cell efficiency (8%), at a fraction of the cost of platinised plastic.
Low temperature fabrication of high performance and transparent Pt counter electrodes for use in flexible dye-sensitized solar cells.
Xiao Y, et al.
Science Bulletin, 57, 2329-2334 (2012)

Questions

  1. What is the ITO coating thickness of product 917311?

    1 answer
    1. The ITO coating thickness of product 917311 is 300 nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service