Skip to Content
Merck
All Photos(2)

Key Documents

916021

Sigma-Aldrich

SHT-263S Solarpur®

greener alternative

Synonym(s):

N2,N2,N2′,N2′,N7,N7,N7′,N7′-octakis(4-methoxyphenyl)-9,9′-spirobi[9H-fluorene]-2,2′,7,7′-tetramine, Spiro-MeOTAD, Spiro-OMeTAD

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$697.00
5 G
$2,590.00

$697.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$697.00
5 G
$2,590.00

About This Item

Empirical Formula (Hill Notation):
C81H68N4O8
CAS Number:
Molecular Weight:
1225.43
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$697.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.5% Residual solvent

mp

247 °C

greener alternative category

SMILES string

N(c%14ccc(cc%14)OC)(c%13ccc(cc%13)OC)c1cc2c(cc1)c3c(cc(cc3)N(c%12ccc(cc%12)OC)c%11ccc(cc%11)OC)C42c5c(ccc(c5)N(c%10ccc(cc%10)OC)c9ccc(cc9)OC)c6c4cc(cc6)N(c8ccc(cc8)OC)c7ccc(cc7)OC

InChI

1S/C81H68N4O8/c1-86-65-29-9-53(10-30-65)82(54-11-31-66(87-2)32-12-54)61-25-45-73-74-46-26-62(83(55-13-33-67(88-3)34-14-55)56-15-35-68(89-4)36-16-56)50-78(74)81(77(73)49-61)79-51-63(84(57-17-37-69(90-5)38-18-57)58-19-39-70(91-6)40-20-58)27-47-75(79)76-48-28-64(52-80(76)81)85(59-21-41-71(92-7)42-22-59)60-23-43-72(93-8)44-24-60/h9-52H,1-8H3

InChI key

XDXWNHPWWKGTKO-UHFFFAOYSA-N

General description

Both SHT-263S and SHT-263 can be offered in bulk quantities.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives, and has been enhanced for energy efficiency. Click here for more information.

Application

  • SHT-263S is a state-of-art, hole-transport material (Spiro-MeOTAD) for perovskite solar cells.
  • SHT-263S offers high purity with modest cost.
  • Together with the highest grade Spiro-MeOTAD, SHT-263 (902500) which has a purity of ≥99.9% (HPLC), these two trade-marked products provide high material purity, low-solvent residual, batch-to-batch consistency, and offer trusted high-performance in devices.

Legal Information

Solarpur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Hybrid Solar Cells Based on Meso-Superstructured Organometal Halide Perovskites
Lee M M, et al.
Science, 338(6107), 643-647 (2012)
Hairen Tan et al.
Science (New York, N.Y.), 355(6326), 722-726 (2017-02-06)
Planar perovskite solar cells (PSCs) made entirely via solution processing at low temperatures (<150°C) offer promise for simple manufacturing, compatibility with flexible substrates, and perovskite-based tandem devices. However, these PSCs require an electron-selective layer that performs well with similar processing.
G Grancini et al.
Nature communications, 8, 15684-15684 (2017-06-02)
Despite the impressive photovoltaic performances with power conversion efficiency beyond 22%, perovskite solar cells are poorly stable under operation, failing by far the market requirements. Various technological approaches have been proposed to overcome the instability problem, which, while delivering appreciable
Julian Burschka et al.
Nature, 499(7458), 316-319 (2013-07-12)
Following pioneering work, solution-processable organic-inorganic hybrid perovskites-such as CH3NH3PbX3 (X = Cl, Br, I)-have attracted attention as light-harvesting materials for mesoscopic solar cells. So far, the perovskite pigment has been deposited in a single step onto mesoporous metal oxide films

Articles

Novel biosensing devices, like bio-FET sensors, offer label-free analysis by detecting ionic and biomolecular charges, crucial for diagnostics and biological research.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service