Skip to Content
Merck
All Photos(1)

Documents

915270

Sigma-Aldrich

Nafion perfluorinated membrane

Nafion 117, thickness 0.007 in., sheet size 7.5 cm × 13.0 cm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7HF13O5S . C2F4)x
CAS Number:
MDL number:
UNSPSC Code:
60104509

form

sheet

manufacturer/tradename

Nafion 117

sheet size

7.5 cm × 13.0 cm

thickness

0.007 in.

solubility

H2O: insoluble

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Nafion membrane may be used in the fabrication of electrodes by dry plasma. As a proton exchange membrane Nafion has been the focus of considerable efforts to develop improved fuel cells.
Nafion is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.
Nafion is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.

Packaging

1 ea in rigid mailer
Darkening/browning is common for pieces stored in a general room environment for an extended period of time (several months), due to organics pick-up. Verify product is kept sealed during storage.
  • Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
  • Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.
Packaged in heat-sealed foil bag in stay-flat mailers
Darkening/browning is common for pieces stored in a general room environment for an extended period of time (several months), due to organics pick-up. Verify product is kept sealed during storage.
· Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
· Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of temperature and humidity on the mechanical properties of Nafion 117 polymer electrolyte membrane.
Bauer F, et al.
Journal of Polymer Science. Part B, Polymer Physics, 43(7), 786 - 795 (2005)
Ion mobility in Nafion-117 membranes
Stenina IA, et al.
Desalination, 170(1), 49-58 (2004)
Evaluation of Adhesion between Nafion and gold electrodes fabricated through novel dry process utilizing argon plasma treatment.
Hiroshiba N, et al.
Chemistry Letters (Jpn), 42(10), 1125-1127 (2013)
Alternative polymer systems for proton exchange membranes (PEMs).
Michael A Hickner et al.
Chemical reviews, 104(10), 4587-4611 (2005-01-27)
Camille Bonomelli et al.
PloS one, 6(8), e23521-e23521 (2011-08-23)
The N-linked oligomannose glycans of HIV gp120 are a target for both microbicide and vaccine design. The extent of cross-clade conservation of HIV oligomannose glycans is therefore a critical consideration for the development of HIV prophylaxes. We measured the oligomannose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service