Skip to Content
Merck
All Photos(1)

Key Documents

914541

Sigma-Aldrich

Molybdenum disulfide quantum dots

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoS2
CAS Number:
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

Quality Level

Looking for similar products? Visit Product Comparison Guide

Application

The transformation of 2D materials in QDs form supplement excellent properties from their quantum confinement effects, surface-volume ratio, solubility, edge-effects, tunable PL, ease to functionalize and fluorescence properties. These 2D QDs are demanding to impart new dimension to the 2D nanosheets and in order to advance to next level, the better understanding of the properties, synthesis methods and applications of 2D QDs are needed. The exciting optical properties of the MoS2 QDs such as strong excitonic absorption bands covering the visible region, excellent excitonic fluorescence, highly tunable PL, atomically-thin structure, molecular size, biocompatibility, easy functionalization, enhanced photostability, higher molar extinction coefficients, and long-term photostability make them attractive candidates for many different applications including energy, batteries, and imaging.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emerging 0D Transition-Metal Dichalcogenides for Sensors, Biomedicine, and Clean Energy
B. Li
Small, 13, 1700527-1700527 (2017)
2D−Materials-Based Quantum Dots: Gateway Towards Next-Generation Optical Devices
S. Dhanabalan
Advanced Optical Materials, 1700257-1700257 (2017)
Research Update: Beyond graphene?Synthesis of functionalized quantum dots of 2D materials and their applications
Musselman, et al.
APL Materials, 6, 120701-120701 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service