Skip to Content
Merck
All Photos(1)

Key Documents

913820

Sigma-Aldrich

Titania nanoparticles

dispersion, 5 wt. % in ethanol, 500 nm particle size

Synonym(s):

TiO2 nanoparticles, TiOxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$219.00

$219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$219.00

About This Item

Linear Formula:
TiO2
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

$219.00


Check Cart for Availability

Request a Bulk Order

form

dispersion
nanoparticles

Quality Level

concentration

5 wt. % in ethanol

particle size

500 nm

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titania nanoparticles are useful for a wide variety of applications, due to their high refractive index and large bandgap, providing optical properties in the visible spectrum. Applications of titania nanoparticles range from energy to medical applications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified titanium oxide (TiO2) nanocomposites and its array of applications: a review.
Ghosh S, et al.
Toxicological & Environmental Chemistry, 97:5, 491-514 (2015)
Introduction: titanium dioxide (TiO2) nanomaterials.
Xiaobo Chen et al.
Chemical reviews, 114(19), 9281-9282 (2014-10-09)
Janusz Bogdan et al.
Nanoscale research letters, 12(1), 225-225 (2017-03-30)
Cancer has become a global problem. On all continents, a great number of people are diagnosed with this disease. In spite of the progress in medical care, cancer still ends fatal for a great number of the ill, either as
Mini review on photocatalysis of titanium dioxide nanoparticles and their solar applications.
Lan Y, et al.
Nano Energy, 2(5), 1031-1045 (2013)

Articles

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Questions

  1. Wenn man einen Durchmesserbereich von 500 nm angibt, sind es dann wirklich 500 nm, oder wie es geschrieben steht, ist es nur der Mittelwert, wenn es sich um den Mittelwert handelt, wie groß der Bereich ist, da ich nach einem kleinen Variationsbereich

    1 answer
    1. The listed 500 nm value is an average. The distribution is defined on the Certificate of Analysis as follows:
      "Mean Particle Size dv90" must be between 450 and 550 nm.

      Please follow the link below to access a Certificate from the DOCUMENTATION section:
      https://www.sigmaaldrich.com/product/aldrich/913820#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service