Skip to Content
Merck
All Photos(2)

Key Documents

913812

Sigma-Aldrich

2-(Azidomethyl)nicotinic acid imidazolide

≥95%

Synonym(s):

2-(Azidomethyl)-3-(1H-imidazole-1-carbonyl)pyridine, NAI-N3, RNA SHAPE probe, icSHAPE reagent

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$278.00
250 MG
$1,060.00

$278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$278.00
250 MG
$1,060.00

About This Item

Empirical Formula (Hill Notation):
C10H8N6O
CAS Number:
Molecular Weight:
228.21
MDL number:
UNSPSC Code:
12352106

$278.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

74-80 °C

storage temp.

−20°C

SMILES string

[N+](=[N-])=NCc1ncccc1C(=O)[n]2cncc2

InChI

1S/C10H8N6O/c11-15-14-6-9-8(2-1-3-13-9)10(17)16-5-4-12-7-16/h1-5,7H,6H2

InChI key

QPSQVTFTPHUCEH-UHFFFAOYSA-N

Application

2-(Azidomethyl)nicotinic acid imidazolide (NAI-N3) is an RNA icSHAPE probe for live-cell RNA structure profiling across the genome. icSHAPE -- or in vivo click selective 2′-hydroxyl acylation and profiling experiment -- uses NAI-N3 in a chemoaffinity method to probe RNA structure. NAI-N3 is an azido version of the cell-permeable SHAPE reagent 2-methylnicotinic acid imidazolide (NAI) that permits the tagging of NAI-N3-modified RNA with a biotin moiety for subsequent capture via streptavidin. Not only do such strategies further the understanding of RNA structure in living cells but also provide a tool for identifying regions that may be susceptible to therapeutic targeting. Recently, the azidomethylnicotinyl (AMN) group of NAI-N3 was demonstrated to block the function of gRNA and CRISPR systems, which could be reactivated by removing the AMN groups with Staudinger reduction (DPBM phosphine), overall providing a means to control nucleic acid cleavage and gene editing in live cells.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Ru Wang et al.
Nature communications, 11(1), 91-91 (2020-01-05)
Prokaryotes use repetitive genomic elements termed CRISPR (clustered regularly interspaced short palindromic repeats) to destroy invading genetic molecules. Although CRISPR systems have been widely used in DNA and RNA technology, certain adverse effects do occur. For example, constitutively active CRISPR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service