Skip to Content
Merck
All Photos(2)

Key Documents

913774

Sigma-Aldrich

IEICO-4Cl

≥99%

Synonym(s):

2,2′-((2Z,2′Z)-(((4,4,9-tris(4-hexylphenyl)-9-(4-pentylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene 5,2-diyl))bis(methanylylidene))bis(5,6-dichloro-3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$531.00

$531.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$531.00

About This Item

Empirical Formula (Hill Notation):
C114H114Cl4N4O4S4
CAS Number:
Molecular Weight:
1874.22
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

$531.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: Eg=1.27 eV

Quality Level

Assay

≥99%

form

solid

color

dark

Orbital energy

HOMO -5.56 eV 
LUMO -4.23 eV 

SMILES string

[s]1c(c(cc1\C=C%14/C(=O)c%15c(cc(c(c%15)Cl)Cl)C/%14=C(C#N)C#N)OCC(CCCC)CC)c2[s]c3c(c2)C(c6c3cc7c(c6)c8[s]c(cc8C7(c%13ccc(cc%13)CCCCCC)c%12ccc(cc%12)CCCCCC)c9[s]c(cc9OCC(CCCC)CC)\C=C%10/C(=O)c%11c(cc(c(c%11)Cl)Cl)C/%10=C(C#N)C#N)(c5ccc(cc5)CCCCCC)c4ccc(cc4

InChI key

SVCAQTPUROMFDI-VMEYAEKKSA-N

Application

IEICO-4Cl is a transparent, low-bandgap non-fullerene acceptor. It is a chlorinated derivative of IEICO, which imparts it with a lower optical bandgap and lower energy levels.[1] The weak photon absorption in the visible spectrum of IEICO-4Cl, allows high-performance organic solar cells to be fabricated with outstanding transparency. Furthermore, changing the polymer acceptor pairing with IEICO-4Cl allows tuning of the color of the solar cell device fabricated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Cui et al.
Advanced materials (Deerfield Beach, Fla.), 29(43) (2017-10-05)
Semitransparent organic solar cells (OSCs) show attractive potential in power-generating windows. However, the development of semitransparent OSCs is lagging behind opaque OSCs. Here, an ultralow-bandgap nonfullerene acceptor, "IEICO-4Cl", is designed and synthesized, whose absorption spectrum is mainly located in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service