Skip to Content
Merck
All Photos(3)

Key Documents

913219

Sigma-Aldrich

(S,S)-BABIBOP

Synonym(s):

(3S,3′S)-3,3′-Di-tert-butyl-2,2′,3,3′-tetrahydro-4,4′-bibenzo[d][1,3]oxaphosphole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O2P2
CAS Number:
Molecular Weight:
386.40
UNSPSC Code:
12352200

form

powder

optical purity

ee: ≥99% (HPLC)

reaction suitability

reagent type: ligand

functional group

phosphine

Application

(S,S)-BABIBOP is a P-chiral biaryl biphosphorus ligand used in Pd, Cu, and Rh-catalyzed asymmetric hydrogenations.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

Sold in collaboration with Zejun Pharmaceuticals

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga V Zatolochnaya et al.
Chemical science, 9(19), 4505-4510 (2018-06-14)
A new class of tunable heterophosphole dimeric ligands have been designed and synthesized. These ligands have enabled the first examples of Cu-catalyzed hydrogenation of 2-substituted-1-tetralones and related heteroaryl ketones via dynamic kinetic resolution, simultaneously creating two contiguous stereogenic centers with
Guisheng Li et al.
Organic letters, 20(7), 1725-1729 (2018-03-16)
Novel bidentate phosphine ligands BABIPhos featuring a biaryl bis-dihydrobenzooxaphosphole core are presented. Their synthesis was achieved via Pd-catalyzed reductive homocoupling of dihydrobenzooxaphosphole aryl triflates. An efficient route toward various analogues was also established, giving access to phosphines with different electronic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service