Skip to Content
Merck
All Photos(2)

Key Documents

913154

Sigma-Aldrich

1,10-Phenanthroline nickel (ll) dibromide

Synonym(s):

(phen)NiBr2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$184.00

$184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$184.00

About This Item

Empirical Formula (Hill Notation):
C12H8Br2N2Ni
CAS Number:
Molecular Weight:
398.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$184.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

Br[Ni]1(Br)[N]2=CC=CC3=C2C([N]1=CC=C4)=C4C=C3

Application

1,10-Phenanthroline nickel (ll) dibromide is a Ni precatalyst used for a variety of cross-coupling reactions, including N-alkylation of amides and arylation of alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jack Twilton et al.
Angewandte Chemie (International ed. in English), 57(19), 5369-5373 (2018-03-01)
The combination of nickel metallaphotoredox catalysis, hydrogen atom transfer catalysis, and a Lewis acid activation mode, has led to the development of an arylation method for the selective functionalization of alcohol α-hydroxy C-H bonds. This approach employs zinc-mediated alcohol deprotonation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service