912573
High surface area conducting polypyrrole
Synonym(s):
3D cross-linked polypyrrole, doped by phytic acid, Doped polypyrrole hydrogels, PPy
Select a Size
About This Item
Recommended Products
description
Electrochemical activity: see attached CV result
Quality Level
form
solid
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
color
black
solubility
soluble (swell into hydrogel but does not dissolve in water)
greener alternative category
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.
Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service