Skip to Content
Merck
All Photos(1)

Key Documents

911631

Sigma-Aldrich

NanoFabTx reagent

Stabilizer-Micro

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$174.00

$174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$174.00

About This Item

Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.51

$174.00


Check Cart for Availability

Request a Bulk Order

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

NanoFabTx reagent, Stabilizer-Micro is a component of our NanoFabTx PLGA-micro formulation kits (912212, 912220), which include additional reagents and protocols for nanoprecipitation and microfluidics to synthesize microparticles 1-30 μm in size. NanoFabTx reagent, Stabilizer-Micro is a polymer stabilizer.

Application

NanoFabTx reagent, Stabilizer-Micro can be used as a refill for the component in our NanoFabTx PLGA-micro formulation kits (912212, 912220).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service