Skip to Content
Merck
All Photos(1)

Key Documents

910740

Sigma-Aldrich

Titanium aluminium carbide 312

≥90%, ≤200 μm particle size

Synonym(s):

MAX Phase 312, Ti3AlC2

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$513.00

$513.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$513.00

About This Item

Linear Formula:
Ti3AlC2
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$513.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90%

form

powder

color

dark gray

particle size

≤200 μm

Looking for similar products? Visit Product Comparison Guide

Application

MAX phases are a family of ternary carbides and nitrides that share a similar layered hexagonal crystal structure. They are so called because of their chemical formula: M(n+1)AXn —where n = 1, 2, or 3, where M is an early transition metal, A is an element from the IIIA or IVA groups, and X is carbon and/or nitrogen class of materials.[1] MAX phases combine attractive properties of both ceramics and metals, and has been suggested for potential applications in nuclear reactor. The specific activities of Ti3AlC2 and Ti2AlC were found to be similar to SiC, and are three orders of magnitude less than Alloy 617 after 10–60 years decay for all three activation times in both the fast and thermal spectra.[2]

MAX phases are important precursors for synthesizing MXene, a highly conductive 2-dimentional nanomaterial. MXenes are produced by selective etching of the A element from the MAX phases. It combine the metallic conductivity of transition metal carbides with the hydrophilic nature of their hydroxyl or oxygen terminated surfaces.[3] Ti3AlC2 MAX phase is one of the most used MAX phase for MXene (Ti3C2Tx).[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guidelines for Synthesis and Processing of Two-Dimensional Titanium Carbide (Ti3C2Tx MXene)
Alhabeb M, et al.
Chemistry of Materials, 29(18), 7633-7644 (2017)
Michael Naguib et al.
Advanced materials (Deerfield Beach, Fla.), 26(7), 992-1005 (2013-12-21)
Recently a new, large family of two-dimensional (2D) early transition metal carbides and carbonitrides, called MXenes, was discovered. MXenes are produced by selective etching of the A element from the MAX phases, which are metallically conductive, layered solids connected by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service