Skip to Content
Merck
All Photos(1)

Key Documents

90976

Sigma-Aldrich

N-Boc-4-(Fmoc-amino)piperidine-4-carboxylic acid

≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N2O6
CAS Number:
Molecular Weight:
466.53
Beilstein:
7609053
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0% (HPLC)

form

powder

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCC(CC1)(NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O

InChI

1S/C26H30N2O6/c1-25(2,3)34-24(32)28-14-12-26(13-15-28,22(29)30)27-23(31)33-16-21-19-10-6-4-8-17(19)18-9-5-7-11-20(18)21/h4-11,21H,12-16H2,1-3H3,(H,27,31)(H,29,30)

InChI key

BOFOACPQHWDRLH-UHFFFAOYSA-N

Other Notes

Amino acid for preparation of water-soluble highly helical peptides[1]
Amino acid for preparing water-soluble highly helical peptides[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-Aminopiperidine-4-carboxylic Acid: A Cyclic alpha,alpha-Disubstituted Amino Acid for Preparation of Water-Soluble Highly Helical Peptides.
Christopher L. Wysong et al.
The Journal of organic chemistry, 61(22), 7650-7651 (1996-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service