Skip to Content
Merck
All Photos(1)

Key Documents

909319

Sigma-Aldrich

2,7-Dibromo-10-methylacridone

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Br2NO
CAS Number:
Molecular Weight:
367.04
UNSPSC Code:
12352101

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

287-291 °C

InChI

1S/C14H9Br2NO/c1-17-12-4-2-8(15)6-10(12)14(18)11-7-9(16)3-5-13(11)17/h2-7H,1H3

InChI key

JVILQLLOKMSADY-UHFFFAOYSA-N

Application

2,7-Dibromo-10-methylacridone is a versatile intermediate for the synthesis of acridinium-based photocatalysts.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service