Skip to Content
Merck
All Photos(1)

Documents

907855

Sigma-Aldrich

(S)-2-((2-Hydroxy-3-methylbenzyl)amino)-N,N,3-trimethylbutanamide

≥95%

Synonym(s):

Hoveyda aminophenol catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
UNSPSC Code:
12352111

Assay

≥95%

form

powder or chunks

mp

58-60 °C

storage temp.

2-8°C

Application

(S)-2-((2-Hydroxy-3-methylbenzyl)amino)-N,N,3-trimethylbutanamide is an aminophenol organocatalyst developed by the Hoveyda lab for enantioselective addition of allylboron compounds to imines, ketones, and other carbonyls, including polyfluorinated substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel W Robbins et al.
Angewandte Chemie (International ed. in English), 55(33), 9610-9614 (2016-06-09)
A set of broadly applicable methods for efficient catalytic additions of easy-to-handle allyl-B(pin) (pin=pinacolato) compounds to ketones and acyclic α-ketoesters was developed. Accordingly, a large array of tertiary alcohols can be obtained in 60 to >98 % yield and up to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service