Skip to Content
Merck
All Photos(2)

Key Documents

906778

Sigma-Aldrich

(2-Isobutyl-2-methyl-1,3-dioxolan-4-yl)methyl acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$196.00
500 ML
$985.00

$196.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$196.00
500 ML
$985.00

About This Item

Empirical Formula (Hill Notation):
C12H20O4
CAS Number:
Molecular Weight:
228.28
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$196.00


Estimated to ship on01 June 2025


Request a Bulk Order

form

liquid

contains

Contains 200 ppm MEHQ as stabilizer

color

white to yellow

storage temp.

2-8°C

SMILES string

C=CC(OCC1OC(C)(CC(C)C)OC1)=O

InChI key

BICNEUKUQARCEL-UHFFFAOYSA-N

Application

(2-Isobutyl-2-methyl-1,3-dioxolan-4-yl)methyl acrylate units lower the Tg of resulting polymers. It can be used for contact lens and medical devices manufacture. Also, it is suitable for 3D printing applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service