Skip to Content
Merck
All Photos(3)

Key Documents

906719

Sigma-Aldrich

Pyrylium tetrafluoroborate

≥95%

Synonym(s):

Pyry-BF4

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$176.00
5 G
$685.00

$176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$176.00
5 G
$685.00

About This Item

Empirical Formula (Hill Notation):
C5H5BF4O
CAS Number:
Molecular Weight:
167.90
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.22

$176.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

[B-](F)(F)(F)F.[o+]1ccccc1

InChI

1S/C5H5O.BF4/c1-2-4-6-5-3-1;2-1(3,4)5/h1-5H;/q+1;-1

InChI key

SJNICXIZTGPXAU-UHFFFAOYSA-N

Application

Pyrylium tetrafluoroborate is used for the functionalization of heterocyclic amines, activating them for nucleophilic substitution. As demonstrated by the Cornella lab, the intermediate pyridinium salts can react with a vast array of nucelophiles to form new C-N, C-O, and C-S bonds, making pyrylium tetrafluoroborate a valuable reagent for late-stage functionalization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Moser et al.
Angewandte Chemie (International ed. in English), 57(34), 11035-11039 (2018-07-04)
The functionalization of aminoheterocycles by using a pyrylium tetrafluoroborate reagent (Pyry-BF4 ) is presented. This reagent efficiently condenses with a great variety of heterocyclic amines and primes the C-N bond for nucleophilic aromatic substitution. More than 60 examples for the

Related Content

The main focus of the Cornella research group is the development of rapid, practical and efficient methodologies for organic synthesis. In addition to efficiency and practicality, the group is highly interested in discovering new reactivity with the aim of unveiling novel transformations. More specifically, on the fundamental understanding and application of catalytic processes and the development of simple reagents. These two approaches have enormous potential and impact across the chemical sciences.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service