Skip to Content
Merck
All Photos(1)

Documents

905682

Sigma-Aldrich

Dimethylsulfoxonium-3-(pyridoyl)methylide

≥95%

Synonym(s):

2-(Dimethyl(oxo)-sulfaneylidene)-1-(pyridin-3-yl)ethan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2S
CAS Number:
Molecular Weight:
197.25
UNSPSC Code:
12352005

Assay

≥95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

133-135 °C

storage temp.

−20°C

SMILES string

O=C(C1=CC=CN=C1)C=S(C)(C)=O

General description

Sulfoxonium ylides are highly stable carbene precursors, serving as a safer alternative to diazo compounds as they that do not generate gases as byproducts. Sulfoxonium ylides have been shown to be effective in a wide array of transition metal catalyzed C-H functionalization and coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gia L Hoang et al.
Tetrahedron, 74(26), 3318-3324 (2018-07-11)
The synthesis of bridgehead N-fused [5,6]-bicyclic heterocycles via rhodium(III)-catalyzed C-H functionalization of C-alkenyl azoles with sulfoxonium ylides is disclosed. Reactions proceeded in good to high yields for a range of aryl, heteroaryl and alkyl sulfoxonium ylides. In addition, 2-alkenyl imidazoles

Related Content

Ketosulfoxonium ylides serve as versatile carbene equivalents for large-scale reactions, releasing dimethyl sulfoxide biproducts.

The Ellman group has participated in the development of a variety of C-H functionalization methods. An electron rich phosphine ligand has proven to be very useful for a variety of Rh(I)-catalyzed C-C bond forming reactions applicable to heterocycle synthesis as exemplified in the recent Science paper “Proton Donor Acidity Controls Selectivity in Nonaromatic Nitrogen Heterocycle Synthesis.” Another useful ligand developed for the highly functional group compatible direct arylation of nitrogen heterocycles is described in a 2008 J. Am. Chem. Soc. paper “Rh(I)-Catalyzed Arylation of Heterocycles via C-H Bond Activation: Expanded Scope through Mechanistic Insight.” The Ellman group also developed the chiral amine reagent tert-Butanesulfinamide, which is extensively used in academics and industry for the asymmetric synthesis of amines. A comprehensive survey of tert-Butanesulfinamide methods and applications up through 2009 is provided in the 2010 Chemical Reviews article, “Synthesis and Applications of tert-Butanesulfinamide.”

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service