Skip to Content
Merck
All Photos(3)

Key Documents

903701

Sigma-Aldrich

tBuDavePhos Pd G3

1:1 THF adduct

Synonym(s):

t-BuDavePhos G3 Palladacycle, t-BuDavePhos Palladacycle

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$285.00

$285.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$285.00

About This Item

Linear Formula:
C35H45N2O3PPdS · C4H8O
UNSPSC Code:
12161600
NACRES:
NA.22

$285.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

CN(C1=C(C2=C(P(C(C)(C)C)C(C)(C)C)C=CC=C2)C=CC=C1)C.NC3=C(C4=C([Pd]OS(C)(=O)=O)C=CC=C4)C=CC=C3.C5COCC5

InChI

1S/C22H32NP.C12H10N.C4H8O.CH4O3S.Pd/c1-21(2,3)24(22(4,5)6)20-16-12-10-14-18(20)17-13-9-11-15-19(17)23(7)8;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-2-4-5-3-1;1-5(2,3)4;/h9-16H,1-8H3;1-6,8-9H,13H2;1-4H2;1H3,(H,2,3,4);/q;;;;+1/p-1

InChI key

PJRGOORCHKDNHB-UHFFFAOYSA-M

General description

tBuDavePhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has a long life in solutions. tBuDavePhos Pd G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Application

tBuDavePhos Pd G3 has been used as a precatalyst for the Synthesis of Aryl Sulfonamides via Palladium-Catalyzed Chlorosulfonylation of Arylboronic Acids. It may also be used for a variety of C-C and C-N cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas C Bruno et al.
Chemical science, 4, 916-920 (2013-05-15)
A series of easily prepared, phosphine-ligated palladium precatalysts based on the 2-aminobiphenyl scaffold have been prepared. The role of the precatalyst-associated labile halide (or pseudohalide) in the formation and stability of the palladacycle has been examined. It was found that
J Robb DeBergh et al.
Journal of the American Chemical Society, 135(29), 10638-10641 (2013-07-11)
A palladium-catalyzed method for the preparation of sulfonamides is described. The process exhibits significant functional group tolerance and allows for the preparation of a number of arylsulfonyl chlorides and sulfonamides under mild conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service