Skip to Content
Merck
All Photos(1)

Documents

901973

Sigma-Aldrich

SunTronic® UV curing dielectric green for screen printing

Synonym(s):

Dielectric ink, SunTronic® DSU4606G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.23

description

Breakdown Voltage (DC): 3100 V/mil
Max. Service Temp.: 175°C
Specific Gravity: 1.26-1.27
Theoretical Coverage: 32-35 m2/kg/mil
Volume Resistivity: >1014 ohm.cm

form

liquid

color

green

viscosity

20-30 poise (Brookfield CAP @25 °C)

storage temp.

2-8°C

General description

DSU4606G is a flexible, green UV curing dielectric designed to be screen printed as an insulating layer for crossovers in membrane switches and a variety of electronic & Printed circuitry constructions.
DSU4606G has excellent adhesion to both treated and untreated PET. It also adheres to a variety of substrates including PC, PI, glass, and ITO.
Drying/Curing

DSU4606G can be cured with a 200-300 Watt/inch (80-120 Watt/cm) medium-pressure Mercury vapor arc lamps, or microwave initiated D-bulb equivalents. Optimum UV dosage should be around 650 mJ/cm2.
Typical Cured Deposits:

196 threads/inch (77 threads/cm): 0.6-0.8 mil

305 threads/inch (120 threads/cm): 0.3-0.5 mil

DSU4606G must be shielded from sunlight or high-intensity light to prevent on-screen curing during prolonged production stoppages.

Cleaning with thinner followed by cleaning with acetone is recommended

Application

DSU4606G is supplied ready for use, but should be lightly stirred before use. It should not require reducing.
DSU4606G is suitable for use on hand; semi-automatic or fully automatic screen printing machines. Polyester or Stainless screens with mesh count 196-305 threads/inch (77-120 threads/cm) can be used depending on desired ink film thickness. Printing two layers is recommended for maximum dielectric properties and to minimize voids. A total cured deposit of 0.75-2.0 mil may be required to meet specific insulation targets.

Legal Information

SunTronic is a registered trademark of Sun Chemical, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

> 110.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service