Skip to Content
Merck
All Photos(1)

Key Documents

901941

Sigma-Aldrich

Thiol poly(ethylene glycol)-block-poly(lactide-co-glycolide)

PEG average Mn 5,000, PLGA average Mn 15,000, lactide:glycolide 50:50

Synonym(s):

Thiol-PEG-b-PLGA, Thiol-PEG-PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$731.00

$731.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$731.00

About This Item

Linear Formula:
C3H7SO(C2H4O)nC2H4O[(C2H2O2)x(C3H4O2)y]m
UNSPSC Code:
51171641
NACRES:
NA.23

$731.00


Check Cart for Availability

Request a Bulk Order

description

Identity: Conforms to structure

form

powder

feed ratio

lactide:glycolide 50:50
lactide:glycolide 50:50±5 (by NMR)

mol wt

PEG Mn 4000-6000  g/mol (by NMR)
PEG average Mn 5,000
PLGA Mn 13500-16500 g/mol (by NMR)
PLGA average Mn 15,000

color

white

storage temp.

−20°C

Application

Thiol poly(ethylene glycol)-block-poly(lactide-co-glycolide) is a thiol-functionalized, biocompatible, amphiphilic block copolymer. Due to its self-assembling properties, this material can be used in the formation of nanoparticles for drug delivery. Additionally, due to the thiol-functionalization of the PEG block, this material can be used to rapidly covalently conjugate biomolecules, API, or targeting ligands. Potential uses include the targeted and/or controlled release of cancer drugs, anti-inflammatory drugs, antibiotics, or anaesthetic agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradable PLGA-b-PEG polymeric nanoparticles: synthesis, properties and nanomedical applications as drug delivery system
Locatelli E, et al
Journal of Nanoparticle Research, 14, 1316-1316 (2012)
Ilaria Monaco et al.
Journal of medicinal chemistry, 60(10), 4510-4516 (2017-05-05)
Polymeric nanoparticles (PNPs) may efficiently deliver in vivo therapeutics to tumors when conjugated to specific targeting agents. Gint4.T aptamer specifically recognizes platelet-derived growth factor receptor β and can cross the blood-brain barrier (BBB). We synthesized Gint4.T-conjugated PNPs able of high
Yihan Xu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1692-1716 (2016-04-22)
Poly (lactic-co-glycolic acid) (PLGA) copolymers have been broadly used in controlled drug release applications. Because these polymers are biodegradable, they provide an attractive option for drug delivery vehicles. There are a variety of material, processing, and physiological factors that impact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service