Skip to Content
Merck
All Photos(1)

Key Documents

901566

Sigma-Aldrich

Propargyl-PEG2-acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$340.00

$340.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$340.00

About This Item

Empirical Formula (Hill Notation):
C8H12O4
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

$340.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: linker

functional group

alkyne
carboxylic acid

storage temp.

−20°C

SMILES string

C#CCOCCOCCC(O)=O

InChI

1S/C8H12O4/c1-2-4-11-6-7-12-5-3-8(9)10/h1H,3-7H2,(H,9,10)

InChI key

NNWHATPXNWOQKD-UHFFFAOYSA-N

Application

Propargyl-PEG2-acid is a heterobifunctional, PEGylated crosslinker featuring a carboxylic acid at one end and propargyl group at the other for reaction with azide-containing compounds using click chemistry. The hydrophillic PEG linker facilitates solubility in biological applications. Propargyl-PEG2-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.
This is a crosslinker with an alkyne group on one end and a carboxyl group on the other end. The alkyne can be used to attach to an azide using copper-catalyzed click chemistry. The compound contains two PEG units to help improve solubility.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service