Skip to Content
Merck
All Photos(1)

Documents

901438

Sigma-Aldrich

Tri-tert-butylphosphine solution

1.0 M in THF

Synonym(s):

P(t-Bu)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27P
CAS Number:
Molecular Weight:
202.32
MDL number:
UNSPSC Code:
12352128

form

liquid

Quality Level

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

concentration

1.0 M in THF

density

0.878 g/mL

functional group

phosphine

SMILES string

CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P/c1-10(2,3)13(11(4,5)6)12(7,8)9/h1-9H3

InChI key

BWHDROKFUHTORW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Valuable ligand for several coupling reactions. This product is offered as the THF solution for more convenient handling.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service