Skip to Content
Merck
All Photos(3)

Key Documents

901415

Sigma-Aldrich

N-(tert-Butyl)-N-((ethoxycarbonothioyl)thio)-3,5-bis(trifluoromethyl)benzamide

Synonym(s):

Alexanian xanthylation reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$226.00

$226.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$226.00

About This Item

Empirical Formula (Hill Notation):
C16H17F6NO2S2
CAS Number:
Molecular Weight:
433.43
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

$226.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mp

40-46 °C

functional group

amine
fluoro

storage temp.

−20°C

SMILES string

FC(F)(F)c1cc(cc(c1)C(=O)N(SC(=S)OCC)C(C)(C)C)C(F)(F)F

InChI key

QDCXZVWZLHPTDJ-UHFFFAOYSA-N

Application

As reported by the Alexanian. laboratory, this reagent is used for site-selective, intermolecular C-H xanthylation of alkanes, leading to the rapid diversification of otherwise inert C-H bonds. Once installed, the xanthate group provides direct access to diverse product analogues via several aliphatic C-H transformations, including halogenation, deuteration, vinylation, and hydroxylation. Product is best stored in foil-wrapped vials in the freezer when not in use; however, it can be weighed out on the bench without risk of decomposition. This product is offered collaboratively with UNC-Chapel Hill and Erik Alexanian.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William L Czaplyski et al.
Journal of the American Chemical Society (2016-10-16)
Intermolecular functionalizations of aliphatic C-H bonds offer unique strategies for the synthesis and late-stage derivatization of complex molecules, but the chemical space accessible remains limited. Herein, we report a transformation significantly expanding the chemotypes accessible via C-H functionalization. The C-H

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service