Skip to Content
Merck
All Photos(3)

Key Documents

901361

Sigma-Aldrich

N-((Methylsulfonyl)oxy)acetimidoyl chloride

≥95%

Synonym(s):

(1-Chloroethylidene)azanyl ester methanesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$132.00

$132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$132.00

About This Item

Empirical Formula (Hill Notation):
C3H6ClNO3S
CAS Number:
Molecular Weight:
171.60
MDL number:
UNSPSC Code:
12352101

$132.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or crystals

functional group

amine
chloro
sulfonic acid

SMILES string

[S](=O)(=O)(ONC(=C)Cl)C

InChI

1S/C3H6ClNO3S/c1-3(4)5-8-9(2,6)7/h5H,1H2,2H3

InChI key

MAVJMVPCGSHQGN-UHFFFAOYSA-N

Application

This air- and moisture-stable reagent enables the direct functionalization of pyridines under mild conditions for late-stage diversification of complex and drug-like molecules.

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick S Fier
Journal of the American Chemical Society, 139(28), 9499-9502 (2017-07-06)
Herein is reported the design and application of a reagent for the direct functionalization of pyridines. These reactions occur under mild conditions and exhibit broad functional group tolerance, enabling the late-stage functionalization of drug-like molecules. The reagent can be easily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service