Skip to Content
Merck
All Photos(4)

Key Documents

901082

Sigma-Aldrich

Carbon nanotube sheet

aligned, size × thickness 100 mm × 100 mm × 1-5 μm

Synonym(s):

MWCNT, MWNT, Multiwall carbon nanotube

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,380.00

$1,380.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,380.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$1,380.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

>95% (carbon)

resistivity

10-1000 Ω/sq

size × thickness

100 mm × 100 mm × 1-5 μm

diameter

10-40 nm (carbon nanotubes)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Multi-walled carbon nanotubes are aligned in a direction. The CNTs are just connected with van der Waals forces, and no binder material is used. The sheet shows high anisotropic transport properties, including electrical conductivity and thermal conductivity.
CNT has a multi-walled structure. Crystal quality of CNT is examined by Raman scattering measurement. Intensity ratio of G-peak (1580 cm-1) and D-peak (1350 cm-1) represents quality of graphitization of carbon materials. G/D of CNT is ∼3, showing high crystal quality. The high crystallinity is supported by transmission electron microscopy.

Preparation Note

Chemical Vapor Deposition (CVD) method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anisotropic carbon nanotube papers fabricated from multiwalled carbon nanotube webs.
Inoue Y, et al.
Carbon, 49(7), 2437-2443 (2011)

Articles

CNT arrays: Vertically aligned nanotubes grown via CVD; drawn into pure, aligned sheets for innovations.

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service