Skip to Content
Merck
All Photos(1)

Key Documents

901055

Sigma-Aldrich

Ethyl 2-bromo-3-thiophenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BrO2S
CAS Number:
Molecular Weight:
235.10
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

Assay

≥95%

form

liquid

storage temp.

2-8°C

InChI

1S/C7H7BrO2S/c1-2-10-7(9)5-3-4-11-6(5)8/h3-4H,2H2,1H3

InChI key

PDYDQPWWVZTVQD-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Kan et al.
Journal of the American Chemical Society, 139(13), 4929-4934 (2017-03-17)
A new nonfullerene small molecule with acceptor-donor-acceptor (A-D-A) structure, namely, NFBDT, based on a heptacyclic benzodi(cyclopentadithiophene) (FBDT) unit using benzo[1,2-b:4,5-b']dithiophene as the core unit, was designed and synthesized. Its absorption ability, energy levels, thermal stability, as well as photovoltaic performances
Design, synthesis, and structural characterization of the first dithienocyclopentacarbazole-based n-type organic semiconductor and its application in non-fullerene polymer solar cells.
Cao Q, et al.
Journal of Material Chemistry A, 5(16), 7451-7461 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service