Skip to Content
Merck
All Photos(1)

Key Documents

900938

Sigma-Aldrich

m-CBP

sublimed

Synonym(s):

3,3′-Di(9H-carbazol-9-yl)-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H24N2
CAS Number:
Molecular Weight:
484.59
MDL number:
UNSPSC Code:
12352103

Assay

>99% (sublimed, HPLC)

form

powder

General description

Phosphorescent host material and TADF emitter in OLEDs.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifan Zhang et al.
Nature communications, 5, 5008-5008 (2014-09-26)
Organic light-emitting diodes are a major driving force of the current information display revolution due to their low power consumption and potentially long operational lifetime. Although electrophosphorescent organic emitters have significantly lower power consumption than fluorescent emitters, the short lifetime
Simple CBP isomers with high triplet energies for highly efficient blue electrophosphorescence.
Gong S, et al.
Journal of Materials Chemistry, 22(7), 2894-2899 (2012)
Twisted Intramolecular Charge Transfer State for Long-Wavelength Thermally Activated Delayed Fluorescence.
Tanaka H, et al.
Chemistry of Materials, 25(18), 3766-3771 (2013)
A novel pyrrolocarbazole donor for stable and highly efficient thermally activated delayed fluorescent emitters.
Kim JH, et al.
Dyes and Pigments, 136, 529-534 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service