Skip to Content
Merck
All Photos(2)

Key Documents

900894

Sigma-Aldrich

Methyltetrazine-PEG4-NHS ester

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31N5O9
CAS Number:
Molecular Weight:
533.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

NHS ester

storage temp.

−20°C

SMILES string

CC1=NN=C(N=N1)C2=CC=C(C=C2)OCCOCCOCCOCCOCCC(ON3C(CCC3=O)=O)=O

Application

Methyltetrazine-PEG4-NHS ester is a crosslinker with enhanced stability and water solubility. This crosslinker has an NHS ester that can be reacted with a primary amine and a methyltetrazine that is reactive with trans-cyclooctenes. Methyltetrazine-PEG4-NHS ester is one of the most stable tetrazines commercially available. In addition to stabilization provided by the electron donating methyl group, the electron donating alkoxy substituent on the aromatic ring further improves the stability of methyltetrazine-PEG4-NHS ester. The aqueous solubility of this reagent is substantially enhanced by a hydrophilic polyethylene glycol (PEG) spacer arm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradable tetra-PEG hydrogels as carriers for a releasable drug delivery system.
Henise J, et al.
Bioconjugate Chemistry, 26(2), 270-278 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service