Skip to Content
Merck
All Photos(2)

Key Documents

900867

Sigma-Aldrich

1-(Bromomethyl)-4-(1,2,2-triphenylethenyl)benzene

Synonym(s):

1-[(4-Bromomethyl)phenyl]-1,2,2-triphenylethene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21Br
CAS Number:
Molecular Weight:
425.36
UNSPSC Code:
12352103

form

powder or crystals

solubility

DMF: soluble
DMSO: soluble
THF: soluble
dichloromethane: soluble

storage temp.

2-8°C

SMILES string

BrCC1=CC=C(C(C2=CC=CC=C2)=C(C3=CC=CC=C3)C4=CC=CC=C4)C=C1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and self-assembly of amphiphilic polymer with aggregation-induced emission characteristics.
Qin AJ, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 55(5), 772-778 (2012)
Haibin Shi et al.
Journal of the American Chemical Society, 134(43), 17972-17981 (2012-10-10)
Real-time monitoring of cell apoptosis could provide valuable insights into early detection of therapy efficiency and evaluation of disease progression. In this work, we designed and synthesized a new live-cell-permeable, fluorescent light-up probe for real-time cell apoptosis imaging. The probe
L-Valine methyl ester-containing tetraphenylethene: aggregation-induced emission, aggregation-induced circular dichroism, circularly polarized luminescence, and helical self-assembly.
Hongkun L, et al.
Materials Horizons, 1(5), 518-521 (2014)
Phenylcarboxyl-decorated tetraphenylethene with diverse molecular RIM-induced emission from host-guest inclusion and aggregation formation.
Yan G, et al.
Royal Society of Chemistry Advances, 6(20), 16581-16585 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service