Skip to Content
Merck
All Photos(2)

Key Documents

900797

Sigma-Aldrich

1,4-Di-tert-butyl-2,5-bis (2-methoxyethoxy)benzene

99.5%, anhydrous

Synonym(s):

DBBB, Redox Shuttle ANL-RS2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
MDL number:
UNSPSC Code:
12352200

grade

anhydrous
battery grade

Assay

99.5%

form

powder

bp

70 °C

application(s)

battery manufacturing

Application

In lithium ion batteries (LIBs) the redox shuttles are employed to prevent overcharging which could lead to thermal runaway and catastrophic failure.[1] The use of redox shuttles increases the safety and stability of LIBs. 1,4-Di-tert-butyl-2,5-bis (2-methoxyethoxy)benzene is an important redox shuttle used for overcharge protection in batteries.[1][2][3]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang L, et al.
ECS Transactions, 45, 57-57 (2013)
Barbarich TJ and Patterson ML
ECS Transactions, 45, 49-49 (2013)
An All-Organic Non-aqueous Lithium-Ion Redox Flow Battery.
Brushett FR, et al.
Advanced Energy Materials, 2, 1390-1396 (2012)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service