Skip to Content
Merck
All Photos(1)

Key Documents

900740

Sigma-Aldrich

Poly(N-isopropylacrylamide) azide terminated

average Mn 5,000, PDI <1.3

Synonym(s):

Azide polyNIPAM, PNIPAAm, PNIPAM-N3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N3(CH2)3OOCC(CH3)2(C6H11NO)nH
UNSPSC Code:
12352200
NACRES:
NA.23

form

powder

mol wt

average Mn 5,000

PDI

<1.3

Looking for similar products? Visit Product Comparison Guide

Application

Poly(N-isopropylacrylamide) (PNIPAAm) is a tunable, stimuli responsive polymer. This material was prepared by controlled radical polymerization to achieve polymer featuring narrow polydispersity and improving reproducibility in applications. The terminal azide functionality can react with acetylene/alkyne groups via click chemistry for facile and rapid functionalization.

Legal Information

Sold for research purposes only. sigma-aldrich.com/raftlicense.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Responsive Polymer-Protein Bioconjugates Prepared by RAFT Polymerization and Copper-Catalyzed Azide-Alkyne Click Chemistry.
Li M, et al.
Macromolecular Rapid Communications, 29, 1172-1176 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service