Skip to Content
Merck
All Photos(1)

Key Documents

900696

Sigma-Aldrich

Graphene dispersion

0.5-1.0 mg/mL in water, with non-ionic surfactant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

dispersion

concentration

0.5-1.0 mg/mL in water

particle size

≤500 nm

bp

100 °C (water)

density

0.9-1.1 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Prepared by the exfoliation of graphite in aqueous solution using Pluronic® F87 as surfactant. Pluronic® F87 is one of nonionic, biocompatible block copolymers.

Legal Information

Sold under Material Transfer Agreement with Mark Hersam group at Northwestern University.
Pluronic is a registered trademark of BASF

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Performance Solid-State Supercapacitors and Microsupercapacitors Derived from Printable Graphene Inks
Lei Li, Ethan B. Secor, Kan-Sheng Chen, Jian Zhu, Xiaolong Liu, Theodore Z. Gao
Advanced Energy Materials, 6, N/A-N/A (2016)
Rapid and Versatile Photonic Annealing of Graphene Inks for Flexible Printed Electronics
Ethan B. Secor , Bok Y. Ahn , Theodore Z. Gao , Jennifer A. Lewis , and Mark C. Hersam
Advanced Materials, 27, 6683-6688 (2015)
Nikhita D Mansukhani et al.
Small (Weinheim an der Bergstrasse, Germany), 12(3), 294-300 (2015-12-01)
Conditions for the dispersion of molybdenum disulfide (MoS2) in aqueous solution at concentrations up to 0.12 mg mL(-1) using a range of nonionic, biocompatible block copolymers (i.e., Pluronics and Tetronics) are identified. Furthermore, the optimal Pluronic dispersant for MoS2 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service