Skip to Content
Merck
All Photos(1)

Documents

900678

Sigma-Aldrich

AMPA-functionalized silica gel

≥99%

Synonym(s):

Aminomethylphosphonic acid-functionalized silica gel, Si-AMPA, Si-Aminopropyl-N1,N-bis (methyl phosphonic acid)-functionalized silica gel, SiliaMetS® AMPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥99%

form

powder

shelf life

2 yr (expiration date period)

reaction suitability

reagent type: catalyst
reagent type: chelator

surface area

480-550 m2/g

capacity

≥0.80 mmol/g loading(molecular loading)

bulk density

707 g/L (typical tap density)

Application

Metal scavenger: Al, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Mn, Nd, Pm, Pr, Sb, Sm, Tb, Tm, V & Yb.

Caution

For prolonged storage, recommended conditions are to store product in a cool place (<8 °C) under inert atmosphere (argon) when possible.

Legal Information

SiliaMetS is a registered trademark of Silicycle, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service