Skip to Content
Merck
All Photos(2)

Key Documents

900555

Sigma-Aldrich

(N-Isocyanoimino)triphenylphosphorane

95%

Synonym(s):

Pinc

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$565.00

$565.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$565.00

About This Item

Empirical Formula (Hill Notation):
C19H15N2P
CAS Number:
Molecular Weight:
302.31
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$565.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

application(s)

peptide synthesis

SMILES string

[P](=N[N+]#[C-])(c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C19H15N2P/c1-20-21-22(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19/h2-16H

InChI key

NIDTXBFHPXMXTR-UHFFFAOYSA-N

General description

(N-Isocyanoimino)triphenylphosphorane (Pinc) is a bench-stable solid reagent widely used in organic synthesis. It is an amphoteric compound with unique properties that make it a versatile tool for various applications, including heterocycle synthesis and macrocyclization of peptides. Pinc facilitates cyclization and incorporation of a conformational control element into peptide macrocycles, leading to desired drug properties such as improved membrane permeability, lipophilicity, and aqueous solubility.[1] Its ability to form oxadiazoles offers opportunities for the development of new transformations.

Application

As reported by the lab of Andrei Yudin, Pinc enables zwitterionic-controlled generation of peptide macrocyles from linear peptides and aldehydes. It enables the formation of disubstituted 1,3,4-oxadiazoles by intercepting the key mixed anhydride characteristic of the Ugi reaction.[2] Furthermore, the resulting peptide macrocycles containing oxadiazole exhibit improved drug properties such as cell-permeability and a rigid conformation.[3]

Features and Benefits

  • Bench-stable solid: Pinc is a stable reagent that can be easily handled and stored.
  • Facilitates cyclization and incorporation of conformational control element: Pinc enables the formation of peptide macrocycles with a desired conformation, leading to improved drug properties.
  • Amphoteric properties: Pinc′s amphoteric nature allows for the design of new multicomponent reactions, expanding its synthetic capabilities.
  • Desired drug properties: The resulting peptide macrocycles exhibit enhanced membrane permeability, lipophilicity, and aqueous solubility, making them desirable for drug development.
  • Versatile transformations: Pinc′s ability to form oxadiazoles opens up opportunities for the development of novel synthetic transformations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclic peptides with heterocycles are cell membrane-permeable : Oxadiazole-containing peptidomimetic macrocycles have potential as drugs
Stu Borman
Chemical and Engineering News, 94(43) (2016)
N-Isocyanoiminotriphenylphosphorane: Synthesis, Coordination Chemistry, and Reactions at the Metal
Weinberger B, Fehlhammer WP.
Angewandte Chemie (International Edition in English), 19(6), 480-481 (1980)
Oxadiazole grafts in peptide macrocycles
Frost JR, Scully CCG, Yudin AK.
Nature Chemistry, 8(12), 1105-1111 (2016)
Novel One-Pot, Four-Component Condensation Reaction: An Efficient Approach for the Synthesis of 2,5-Disubstituted 1,3,4-Oxadiazole Derivatives by a Ugi-4CR/aza-Wittig Sequence
Ramazani A, Rezaei A.
Organic Letters, 12(12), 2852-2855 (2010)

Articles

Isocyanides are widely used reagents in organic synthesis, with applications ranging from materials science to drug discovery.

Related Content

The Yudin laboratory is known for the development of amphoteric molecules and their application in synthesis. The corresponding reagents possess nucleophilic and electrophilic functional groups that do not prematurely react with each other.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service