Skip to Content
Merck
All Photos(2)

Key Documents

900542

Sigma-Aldrich

(2S)-2-Phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole

95%

Synonym(s):

Birman (S)-HBTM Resolution Catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$236.00
1 G
$1,210.00

$236.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$236.00
1 G
$1,210.00

About This Item

Empirical Formula (Hill Notation):
C16H14N2S
CAS Number:
Molecular Weight:
266.36
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$236.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

95%

form

powder or chunks

mp

125 °C

functional group

phenyl
thioether

storage temp.

2-8°C

SMILES string

C12=CC=CC=C1SC3=N[C@H](C4=CC=CC=C4)CCN23

InChI

1S/C16H14N2S/c1-2-6-12(7-3-1)13-10-11-18-14-8-4-5-9-15(14)19-16(18)17-13/h1-9,13H,10-11H2/t13-/m0/s1

InChI key

ZMYZJAQMQBHNLH-ZDUSSCGKSA-N

Application

Organocatalyst for the kinetic resolution of secondary alcohols by acylation with anhydrides.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ximin Li et al.
The Journal of organic chemistry, 77(4), 1722-1737 (2012-01-31)
Kinetic resolution of racemic alcohols has been traditionally achieved via enzymatic enantioselective esterification and ester hydrolysis. However, there has long been considerable interest in devising nonenzymatic alternative methods for this transformation. Amidine-based catalysts (ABCs), a new class of enantioselective acyl

Related Content

The main focus of research in the Birman group is on the de novo design of asymmetric catalysts and reagents. As part of this effort, they have developed Amidine-Based Catalysts, or ABCs, and demonstrated their high enantioselectivity in many asymmetric acyl transfer reactions. The versatility, accessibility, and ease of structural modification of ABCs have attracted the interest of a number of other research groups worldwide, which has led to further expansion of their synthetic utility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service