Skip to Content
Merck
All Photos(2)

Key Documents

900440

Sigma-Aldrich

Orange light-emitting PPV copolymer

Synonym(s):

Livilux® PDO-123

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,900.00

$1,900.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$1,900.00

About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

$1,900.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mol wt

average Mn 340,000
average Mw 1,750,000

Application

Features:
  • Soluble orange light-emitting PPV copolymer.
  • Excellent reproducibility.
  • Broad emission spectrum.
  • High efficiency.
  • Very low operation voltage.
  • Very long DC and AC lifetime.
  • High temperature stability (>100 °C).

Typical performance measured in standard device stack (ITO/PEDOT:PSS 20 nm/PDO-123 80 nm/Ba 6 nm/Ag 150 nm):
  • Max. efficiency: 5.0 Cd/A.
  • V_on: 2.0 V.
  • V at 100 cd/m2: 2.6 V.
  • V at 1000 cd/m2: 3.3 V.
  • Max. power eff.: 5.5 lm/W.
  • External quantum eff.: 3.5%.
  • CIE1931 at 100 cd/m2: (0.60, 0.40).
  • DC lifetime at 100 cd/m2 at RT: >>25000.

Preparation Note

  • 5 g/L in toluene for spin-coating.
  • Special formulation needed for Inkjet.
  • Preparation of device should be in N2 atmosphere.

Legal Information

Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany
Livilux is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service