Skip to Content
Merck
All Photos(1)

Key Documents

900398

Sigma-Aldrich

SnAP-ex 3-N-Boc P Reagent

95%

Synonym(s):

tert-Butyl (4-amino-1-(tributylstannyl)butyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$307.00

$307.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$307.00

About This Item

Empirical Formula (Hill Notation):
C21H46N2O2Sn
Molecular Weight:
477.31
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$307.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

paste

refractive index

n/D 1.4963

density

1.1316 g/mL

functional group

amine

storage temp.

−20°C

Application

SnAP reagents provide a one-step route, in tandem with various aldehyde substrates, to saturated N-heterocycles. The synthesis of N-heterocycles through SnAP reagents requires mild reaction conditions, and aldehydes bearing aryl, heteroaryl, glyoxyl, aliphatic, and halogenated groups are well tolerated. This product was introduced in collaboration with the Bode research group.

related product

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SnAP-eX reagents for the synthesis of exocyclic 3-amino-and 3-alkoxypyrrolidines and piperidines from aldehydes.
Luescher M U and Bode J W
Organic Letters, 18(11), 2652-2655 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service