Skip to Content
Merck
All Photos(1)

Documents

900396

Sigma-Aldrich

Furfuryl mercaptan-(methylene-d2)

≥97 atom % D, ≥97% (CP)

Synonym(s):

2-Furanmethane-α,α-d2 thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D2H4OS
CAS Number:
Molecular Weight:
116.18
MDL number:
UNSPSC Code:
12352005

isotopic purity

≥97 atom % D

Assay

≥97% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]C(S)([2H])C1=CC=CO1

InChI

1S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7H,4H2/i4D2

InChI key

ZFFTZDQKIXPDAF-APZFVMQVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F

Flash Point(C)

45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service