Skip to Content
Merck
All Photos(1)

Key Documents

900386

Sigma-Aldrich

2′-Deoxyadenosine-13C10,15N5 5′-monophosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)

Synonym(s):

2′-Deoxy-5′-adenylic-13C10,15N5 acid sodium salt solution, dAMP-13C10,15N5 disodium salt solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$198.00
10 MG
$749.00
25 MG
$1,340.00

$198.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$198.00
10 MG
$749.00
25 MG
$1,340.00

About This Item

Empirical Formula (Hill Notation):
13C10H1215N5Na2O6P
Molecular Weight:
390.08
UNSPSC Code:
41106305
NACRES:
NA.12

$198.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom %

Assay

≥95% (CP)

form

solution

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+15

storage temp.

−20°C

SMILES string

O[13C@H]1[13CH2][13C@H]([15N]2[13CH]=[15N][13C]3=[13C]2[15N]=[13CH][15N]=[13C]3[15NH2])O[13C@@H]1[13CH2]OP(O[Na])(O[Na])=O

Related Categories

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoyan Nie et al.
Reproductive toxicology (Elmsford, N.Y.), 95, 66-74 (2020-05-25)
Cyclophosphamide (Cy), a chemotherapeutic agent, is widely used to treat tumoursand is also associated with premature ovarian insufficiency. 4-Hydroperoxycyclophosphamide (4-HC), an active metabolite of Cy, was used for in vitro experiments. Granulosa cells (GCs) are crucial for maintaining follicle development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service