Skip to Content
Merck
All Photos(1)

Documents

900295

Sigma-Aldrich

Poly(L-lactide)

viscosity 1.5 dL/g 

Synonym(s):

PLA, PLLA, Polylactide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C3H4O2)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips or granules (chunks)

viscosity

1.5 dL/g

transition temp

Tm 167-175 °C
Tg 57 °C

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service