Skip to Content
Merck
All Photos(2)

Key Documents

900291

Sigma-Aldrich

Poly(caprolactone-co-glycolide)

caprolactone:glycolide 95:5, viscosity 1.0 dL/g 

Synonym(s):

PCL-PGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H10O2)m(C4H4O4)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder or granules (chips)

feed ratio

caprolactone:glycolide 95:5

viscosity

1.0 dL/g

transition temp

flash point >230 °F
Tm 53-57 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service