Skip to Content
Merck
All Photos(1)

Key Documents

900170

Sigma-Aldrich

Cortisone-2,2,4,6,6,9,12,12-d8

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$666.00
5 MG
$1,680.00

$666.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$666.00
5 MG
$1,680.00

About This Item

Empirical Formula (Hill Notation):
C21D8H20O5
CAS Number:
Molecular Weight:
368.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

$666.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+8

storage temp.

−20°C

SMILES string

OCC([C@@]1(O)CC[C@@]([C@]1(C)C2([2H])[2H])([H])[C@]3([H])CC([2H])([2H])C4=C([2H])C(C([2H])([2H])C[C@]4(C)[C@@]3([2H])C2=O)=O)=O

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-15,18,22,26H,3-8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1/i3D2,5D2,9D,10D2,18D

InChI key

MFYSYFVPBJMHGN-BMUVRMCQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cortisone-2,2,4,6,6,9,12,12-d8 (Cortisone-d8) is deuterium-labeled Cortisone. Cortisone is an oxidized metabolite of cortisol (glucocorticoid).

Application

Cortisone-2,2,4,6,6,9,12,12-d8 is used as an isotope.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service