Skip to Content
Merck
All Photos(1)

Documents

900066

Sigma-Aldrich

2-Methylpropane-1-13C/Argon Gas Mixture

ratio (1:99), ≥99 atom % 13C

Synonym(s):

Isobutane-1-13C/Argon (RG) Gas Mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352001

isotopic purity

≥99 atom % 13C

form

gas

composition

ratio (1:99)

mass shift

M+1

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service