Skip to Content
Merck
All Photos(1)

Key Documents

89660

Sigma-Aldrich

o-Tolunitrile

≥97.0% (GC)

Synonym(s):

2-Methylbenzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$139.00

$139.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$139.00

About This Item

Linear Formula:
CH3C6H4CN
CAS Number:
Molecular Weight:
117.15
Beilstein:
1857417
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$139.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.5279 (lit.)
n20/D 1.528

bp

205 °C (lit.)

mp

−13-−11 °C (lit.)
−13-−11 °C

density

0.989 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

Cc1ccccc1C#N

InChI

1S/C8H7N/c1-7-4-2-3-5-8(7)6-9/h2-5H,1H3

InChI key

NWPNXBQSRGKSJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Arturo Ruiz-Santoyo et al.
The Journal of chemical physics, 144(4), 044303-044303 (2016-02-02)
Rotationally resolved fluorescence excitation spectra of the origin bands in the S1 ← S0 transition of 2-tolunitrile (2TN) and 3-tolunitrile (3TN) have been recorded in the collision-free environment of a molecular beam. Analyses of these data provide the rotational constants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service